Learn More
Keystone Cleanroom Products Sterile Zip Lock Bag with Slide Closure
Sfoglia Tutto Keystone Cleanroom Products Prodotti
Click to view available options
Lunghezza (sistema imperiale):
12 in.
15 in.
N. per confezione:
2 Pk.
20 Pk.
Dimensione della confezione:
pezzi 250
pezzi 400
pezzi 500
Descrizione
- Pre-sterilized and packaged in multiple layers of Tyvek peel pouches and poly bags to gain access into the aseptic area without excessive spraying and wiping with sanitisation agents
- The bags are used for the removal and transport of media plates from the aseptic area to QC laboratories
- Manufactured and packaged in an ISO certified cleanroom
- Tear and puncture resistant 2.7 mill low density polyethylene
- Each bag has an integral zip seal with the slide lock closure system
- Two zip-lock bags are double packaged in Tyvek/film chevron peel pouches and then inside a third, sealed poly bag layer
- Labelled with lot number, date of manufacture and expiration date
- Validated sterile to a 10-6 SAL
- Certificates of conformance and sterility sent with each shipment
Specifica
Specifica
| Da utilizzare con (applicazione) | Transport EM Media Plates |
| Materiale | LDPE |
| Sterilità | Sterile |
| Senza lattice | Yes |
| N. per confezione | 20 Pk. |
| Metodo di sterilizzazione | Gamma |
| Spessore (sistema imperiale) | 2.7 mil |
| Lunghezza (sistema imperiale) | 15 in. |
| Lunghezza (sistema metrico) | 38.1 cm |
| Larghezza (sistema imperiale) | 12 in. |
| Vedi altri risultati |