Learn More
Greiner Bio-One VIEWseal Adhesive Microplate Sealer
Click to view available options
Adesivo:
Silicone
Dimensione della confezione:
pezzi 1200
Descrizione
- Coated with silicone adhesive
- Non-Pyrogenic
- Highly transparent film for optical measurements
- Sticks only when film surface is pressed on
- Film does not adhere to gloves
- Temperature-resistant from -80°C to
Specifica
Specifica
| Colore | Transparent |
| Sterilità | Non-sterile |
| N. per custodia | 100 |
| Product Type | Sealer |
| Adesivo | Silicone |
| Plate Compatibility | Microplate |
For research use only. Not for use in diagnostic procedures, unless otherwise indicated.