Learn More
3M™ Vinyl Tape 471F
Codice prodotto. 17287770
Click to view available options
Colore:
Black
Blue
Green
Red
White
Yellow
Dimensione della confezione:
24 rotoli
Codice prodotto. 17287770
Marca: 3M™ 7000028853
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Commonly used for marking hazards and aisles, color-coding, paint masking, decorative trim, splicing, and more. Made from vinyl backing with rubber adhesive, our versatile tape resists scrapes, wear, and weathering for long lasting use.
  • Maintains its color when exposed to abrasion, wear and many solvents
  • Unique stretch properties enable tape to remain stretched to conform to uneven surfaces without lifting and retracting
  • Ideal for use on curved and irregular surfaces
  • Easy to apply, excellent holding strength on many surfaces
  • Available in many distinctive colours (yellow, white, red, black, brown, green, orange, purple, blue and transparent)
  • Available in a wide variety of vivid colours and sizes
TRUSTED_SUSTAINABILITY
Specifica
Dimensioni (L x P) 50 mm x 33 m
Altezza (sistema metrico) 0.1 mm
Lunghezza (sistema metrico) 32.9 m
Spessore 0.14 mm
Larghezza (sistema metrico) 5.1 cm