Learn More
Vinylacetic acid, 96%
Codice prodotto. p-7044636
Click to view available options
Quantità:
25 g
100 g
Dimensione della confezione:
100g
25g
Codice prodotto. 11331947
Marca: Thermo Scientific Alfa Aesar L07557.14
Richiesta di formato massivo o personalizzato
CAS: 625-38-7 | C4H6O2 | 86.09 g/mol
Vinylacetic acid used in the preparation of strained nitroso Diels-Aldrer adducts which undergo a domino ROC metathesis in the presence of Ru-carbene catalysts and olefins. It is used in the synthesis of polyethylene glycolated boltorn coatings. It was used to study the inhibition of the enzyme-catalyzed conversion of D-tyrosyl- phenylalanyl-glycine to D-tyrosyl-phenylaline amide by non-peptide carboxylic acids.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Vinylacetic acid used in the preparation of strained nitroso Diels-Aldrer adducts which undergo a domino ROC metathesis in the presence of Ru-carbene catalysts and olefins. It is used in the synthesis of polyethylene glycolated boltorn coatings. It was used to study the inhibition of the enzyme-catalyzed conversion of D-tyrosyl- phenylalanyl-glycine to D-tyrosyl-phenylaline amide by non-peptide carboxylic acids.
Solubility
Fully miscible.
Notes
Store at 4°C. Protect from heat. Store away from oxidizing agents. Incompatible with oxidizing agents, heat, Bases, Reducing agents.
TRUSTED_SUSTAINABILITY
Identificatori chimici
CAS 625-38-7
Formula molecolare C4H6O2
Molecular Weight (g/mol) 86.09
Numero MDL MFCD00002782
InChI Key PVEOYINWKBTPIZ-UHFFFAOYSA-N
Sinonimo 3-butenoic acid, vinylacetic acid, vinyl acetic acid, acetic acid, ethenyl, beta-butenoic acid, .beta.-butenoic acid, 2-propenylcarboxylic acid, allylic acid, vinylessigsaeure, 3-butenoicacid
PubChem CID 32743
ChEBI CHEBI:35897
IUPAC Name but-3-enoic acid
SMILES OC(=O)CC=C
Specifica
Materiale o nome chimico Vinylacetic acid
Punto di fusione -39°C
Densità 1.012
Punti di ebollizione 162°C to 163°C
Punto d'infiammabilità 65°C (149°F)
Formula lineare H2C=CHCH2CO2H
Indice di rifrazione 1.424
Quantità 25 g
Numero UN UN3265
Beilstein 1699159
Informazioni di solubilità Fully miscible.
Peso formulazione 86.09
Percent Purity 96%