missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Viral Dengue Virus 2 NS1 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Marca: R&D Systems MAB94391-SP
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Dengue Virus 2 NS1 Monoclonal specifically detects Dengue Virus 2 NS1 in Viral samples. It is validated for Western Blot.
Specifica
| Dengue virus 2 NS1 | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 5 ug/mL | |
| NP_739584 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Detects recombinant Dengue Virus 2 NS1 protein in direct ELISAs and Western blots. In Direct Elisa and Western Blots, no cross-reactivity with recombinant Dengue Virus 1, 3 and 4 NS1 proteins was observed. | |
| Virus | |
| Purified |
| Western Blot | |
| 969828 | |
| Unconjugated | |
| Lyophilized from a 0.2 um filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 um filtered solution in PBS. with No Preservative | |
| DEN-2, Dengue NS1, Dengue NS1 protein, Dengue Virus, Dengue virus Serotype 2, Dengue virus Type 2, DENV2, DENV2-NS1 | |
| Human embryonic kidney cell line HEK293-derived recombinant Dengue Virus 2 NS1 protein Asp1-Ala352 Accession No. NP_739584 | |
| 25 ug | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
RUO
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto