Learn More
Viral HIV-1 Gag p24 Antibody, R&D Systems™
Rabbit Monoclonal Antibody
Marca: R&D Systems MAB9539-SP
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
HIV-1 Gag p24 Monoclonal specifically detects HIV-1 Gag p24 in Viral samples. It is validated for Western Blot.
Specifica
| HIV-1 Gag p24 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 155030 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG |
| Western Blot | |
| 2230C | |
| Western Blot 0.1 ug/mL | |
| Q77YG1 | |
| E. coli-derived recombinant viral HIV-1 Gag p24 Pro149-Gly354 Accession # Q77YG1 | |
| 25 μg | |
| Primary | |
| Detects viral HIV-1 Gag p24 in Western blots. | |
| Virus | |
| Purified |