missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Viral Tick-borne Encephalitis Virus NS1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Marca: R&D Systems MAB101061-100
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Tick-borne Encephalitis Virus NS1 Monoclonal specifically detects Tick-borne Encephalitis Virus NS1 in Viral samples. It is validated for Western Blot, ELISA.
Specifica
| Tick-borne Encephalitis Virus Envelope | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| Human embryonic kidney cell line HEK293-derived viral Tick-borne Encephalitis Virus NS1, Amino Acids 777-1128 | |
| 100 μg | |
| Detects Viral Tick-borne Encephalitis Virus NS1 in direct ELISAs. | |
| Virus | |
| Ascites |
| Western Blot, ELISA | |
| 1004120 | |
| Western Blot 1 μg/mL, ELISA | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
| IgG2b |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto