missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VISTA Mouse anti-Human , PE-CF594, Clone: MIH65.rMAb (also known as MIH65), BD Biosciences™
Mouse Monoclonal Antibody
Marca: BD Biosciences 566773
Dettagli aggiuntivi : Peso : 0.00020kg
Descrizione
VISTA Monoclonal antibody specifically detects VISTA in Human samples. It is validated for Flow CytometrySpecifica
VISTA | |
Monoclonal | |
PE-CF594 | |
VSIR; B7-H5; B7H5; C10orf54; DD1alpha; GI24; SISP1 | |
Human VISTA Transfected Cell Line | |
RUO | |
Human | |
IgG1 κ |
Flow Cytometry | |
MIH65.rMAb (also known as MIH65) | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Mouse | |
25 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ PE-CF594 under optimum conditions, and unconjugated antibody and free PE-CF594 were removed. |