Learn More
Ismatec™ Viton™ Microbore 2-Stop Tube Sets
Click to view available options
Diametro (sistema metrico) interno:
0.89 mm
1.42 mm
2.79 mm
Portata:
130 mL/min
200 mL/min
30 mL/min
67 mL/min
Dimensione della confezione:
pezzi 12
Descrizione
- Genuine Ismatec™ tubing undergoes robust testing to provide optimal performance/pump life hours with peristaltic pump equipment
- Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
- Global market leader in range of formulations for most extensive chemical compatibility
- Single-use custom lengths and configurations available to minimize downtime
Specifica
Specifica
| Certificazione/Conformità | FDA approved; Prop 65 NA (no Prop 65 chemicals) |
| Diametro (sistema metrico) interno | 1.42 mm |
| Materiale | Viton |
| Overall Length (English) | 1.25 ft. |
| Overall Length (Metric) | 0.38 m |
| Da utilizzare con | Pump Series L/S |
| Min. Operating Temperature (Metric) | 0°C |
| Quantità confezione | 12 / Pk. |
| Portata | 67 mL/min |
| Pressione massima | 25.3 psi, 1.75 bar |
| Vedi altri risultati |