Learn More
Novus Biologicals™ Von Willebrand Factor Antibody (INSERM patent anti-vWF) - VHH-Fc, Novus Biologicals™
Human Monoclonal Antibody
Marca: Novus Biologicals™ NBP3-28369-1mg
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Von Willebrand Factor Monoclonal antibody specifically detects Von Willebrand Factor in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifica
| Von Willebrand Factor | |
| Monoclonal | |
| Unconjugated | |
| P04275 | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| INSERM patent anti-vWF | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| coagulation factor VIII VWF, F8, F8VWF, von Willebrand factor, VWD, vWF | |
| vWF | |
| 1 mg | |
| Cancer | |
| 7450 | |
| Human | |
| Purified |