Learn More
Grant Instruments™ VR Rack for Grant Heated Circulating Bath
Codice prodotto. 11436477
Click to view available options
Capacità:
(102) 0.5 mL Tubes
(14) 30 mm Tubes
(23) 24 mm Tubes
(36) 16 to 19 mm Tubes
(65) 10 to 13 mm Tubes
(75) 1.5 mL Tubes
Dimensione della confezione:
Pezzo
Codice prodotto. 11436477
Marca: Grant Instruments™ VR13
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
For use with a number of Grant Heating Circulators and Heated Circulating Baths. Grant Instruments™ VR Racks for Grant Heated Circulating Baths accommodate a variety of tube sizes and capacities.
  • Accommodates tubes as small as 0.5mL or as large as 30mm dia.
  • For use with Grant Heated Circulating Baths and Heating Circulators.
TRUSTED_SUSTAINABILITY
Specifica
Da utilizzare con (apparecchiatura) T100, TC120, TX150, TXF200 Heating Circulators
Capacità (65) 10 to 13 mm Tubes