Learn More
Grant Instruments™ VR Rack for Grant Heated Circulating Bath
Click to view available options
Capacità:
(102) 0.5 mL Tubes
(14) 30 mm Tubes
(23) 24 mm Tubes
(36) 16 to 19 mm Tubes
(65) 10 to 13 mm Tubes
(75) 1.5 mL Tubes
Dimensione della confezione:
Pezzo
Descrizione
For use with a number of Grant Heating Circulators and Heated Circulating Baths. Grant Instruments™ VR Racks for Grant Heated Circulating Baths accommodate a variety of tube sizes and capacities.
- Accommodates tubes as small as 0.5mL or as large as 30mm dia.
- For use with Grant Heated Circulating Baths and Heating Circulators.
Specifica
Specifica
| Da utilizzare con (apparecchiatura) | T100, TC120, TX150, TXF200 Heating Circulators |
| Capacità | (75) 1.5 mL Tubes |