Learn More
Thermo Scientific™ Water Purification Systems Storage Reservoirs
Codice prodotto. 11367061 Sfoglia Tutto Thermo Scientific Prodotti
Click to view available options
Tipo:
Capacità 60l
Feed Water Tank
Sterile Overflow
Storage Reservoir
Capacità (sistema metrico):
100 L
30 l
30 to 100 L
30 l
60 L
60 l
Include:
Level Display
Level Display, Recirculation Pump
N/A
Reservoir
Tank
Da utilizzare con (apparecchiatura):
GenPure, GenPure Pro, GenPure xCAD
Pacific RO, Pacific TII
Pacific TII
Smart2Pure
Smart2Pure 12
Smart2Pure, Pacific and LabTower Water Purification Systems
Dimensione della confezione:
Pezzo
Codice prodotto. 11367061
Marca: Thermo Scientific™ 50132714
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
I serbatoi compatibili con i sistemi Barnstead™ GenPure™, Pacific™ o Smart2Pure™ Thermo Scientific™ consentono di conservare in sicurezza l'acqua purificata.
Safely store purified water from a compatible system with Thermo Scientific™ Water Purification Systems Storage Reservoirs. For use with Thermo Scientific™ Barnstead™ GenPure™, Pacific™ or Smart2Pure™ systems, these reservoirs are compatible with the following models: GenPure, GenPure Pro, GenPure xCAD, GenPure xCAD Plus, Pacific RO, Pacific TII and Smart2Pure.
Features:
  • Large opening offers easy and effective cleaning.
  • Optional safety filters include a CO2 absorber prevents conductivity increases from drawn-in CO2 and a sterile overflow to prevent microorganisms from entering and contaminating reservoir water.
  • Polyethylene reservoir is constructed with inert materials opaque to light.
  • Recirculation pump (optional) protects water from bacterial growth during standstill while maintaining a low conductivity value.
  • Wall mount is optional.
  • Conical bottom outlet allows for complete draining.
For use with GenPure, GenPure Pro, and GenPure xCAD purification systems only:
  • 06.5038
  • 06.5068
For use with Pacific RO water purification systems only:
  • 06.5034
  • 06.5064
  • 06.5084
For use with Pacific TII water purification systems only:
  • 06.5032
  • 06.5062
  • 06.5082
For use with Pacific RO & Pacific TII water purification systems only:
  • 06.5033
  • 06.5063
  • 06.5083
For use with Smart2Pure 12 water purification systems only:
  • 06.5040
  • 06.5070
TRUSTED_SUSTAINABILITY
Specifica
Capacità (sistema metrico) 30 to 100 L
Da utilizzare con (apparecchiatura) Smart2Pure, Pacific and LabTower Water Purification Systems
Tipo Sterile Overflow
Include N/A