Learn More
Nabertherm™ Weighing Furnace, 1100 LT Series
Codice prodotto. 17338019 Sfoglia Tutto Nabertherm Prodotti
Click to view available options
Tipo di unità di controllo:
B510
C550
Dimensione della confezione:
Pezzo
Codice prodotto. 17338019
Marca: Nabertherm™ LW095H1ANL
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Designed especially for combustion loss determination in the laboratory. The included software records both the temperature and the weight loss.
  • Maximum Temperature: 1100°C
  • Heating from two sides
  • Ceramic heating plates with integral heating element which is safeguarded against fumes and splashing, and easy to replace
  • Only fiber materials are used which are not classified as carcinogenic according to TRGS 905, class 1 or 2
  • Dual shell housing made of sheets of textured stainless steel
  • Optional lift door with hot surface facing away from the operator
  • Adjustable working air inlet in the door
  • Exhaust air outlet in rear wall of furnace
  • Solid state relays provide for low noise operation
  • Delivery includes base, ceramic plunger with base plate in the furnace lining, precision scale and software package
  • 4 scales available for different maximum weights and scaling ranges
  • Process control and documentation for temperature and combustion loss via VCD software package for monitoring, documentation and control
  • Defined application within the constraints of the operating instructions
Specifica
Tipo di unità di controllo B510
Capacità (sistema metrico) 9 L
Temperatura (sistema metrico) 1100°C
Dimensioni interne (L x P x A) 240 x 230 x 170 mm
Dimensioni esterne (L x P x A) 455 x 415 x 980 mm
Peso (sistema metrico) 35 kg