Learn More
Nabertherm™ Weighing Furnace, 1100 LT Series
Sfoglia Tutto Nabertherm Prodotti
Click to view available options
Tipo di unità di controllo:
B510
C550
Dimensione della confezione:
Pezzo
Descrizione
- Maximum Temperature: 1100°C
- Heating from two sides
- Ceramic heating plates with integral heating element which is safeguarded against fumes and splashing, and easy to replace
- Only fiber materials are used which are not classified as carcinogenic according to TRGS 905, class 1 or 2
- Dual shell housing made of sheets of textured stainless steel
- Optional lift door with hot surface facing away from the operator
- Adjustable working air inlet in the door
- Exhaust air outlet in rear wall of furnace
- Solid state relays provide for low noise operation
- Delivery includes base, ceramic plunger with base plate in the furnace lining, precision scale and software package
- 4 scales available for different maximum weights and scaling ranges
- Process control and documentation for temperature and combustion loss via VCD software package for monitoring, documentation and control
- Defined application within the constraints of the operating instructions
Specifica
Specifica
| Tipo di unità di controllo | C550 |
| Capacità (sistema metrico) | 9 L |
| Temperatura (sistema metrico) | 1100°C |
| Dimensioni interne (L x P x A) | 240 x 230 x 170 mm |
| Dimensioni esterne (L x P x A) | 455 x 415 x 980 mm |
| Peso (sistema metrico) | 35 kg |