missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ West Nile Virus Envelope (Domain III) (lineage 1, strain NY99) Monoclonal Antibody (15)

Mouse Monoclonal Antibody
Marca: Invitrogen™ MA536036
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
West Nile Virus Envelope (Domain III) (lineage 1, strain NY99) Monoclonal Antibody for Western Blot
West Nile Virus (WNV) is a member of the Flaviviridae, a plus-stranded virus family that includes St. Louis encephalitis virus, yellow fever virus, and Dengue virus. WNV was initially isolated in 1937 in the West Nile region of Uganda and has become prevalent in Africa, Asia, and Europe. It has rapidly spread across the United States with cases being observed in every continental state. Virus particles consist of a dense core made up of the core/capsid protein encapsulating the RNA genome surrounded by a membrane envelope embedded with envelope and matrix proteins which play a major role for WNV entry into target cells. The viral core protein is thought to contribute to the WNV-associated inflammation via apoptosis induced though the caspase-9 pathway as delivery of core gene delivery into the striatum of mouse brain and skeletal muscle resulted in cell death and inflammation.
Specifica
| West Nile Virus Envelope (Domain III) (lineage 1, strain NY99) | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| Mouse | |
| Protein A | |
| RUO | |
| Virus | |
| Antibody | |
| IgG2a |
| Western Blot | |
| 15 | |
| Unconjugated | |
| West Nile envelope; West Nile Virus; WNV envelope | |
| Recombinant West Nile Virus (WNV) (lineage 1, strain NY99) E/Envelope Protein (Domain III) Protein. | |
| 100 μL | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto