Learn More
Cytiva Whatman™ Ashless Clippings
Codice prodotto. 11909347
Click to view available options
Quantità:
500 g
Dimensione della confezione:
500g
Codice prodotto. 11909347
Marca: Cytiva 1703050
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Enhance filtration speed by coagulating precipitates or suspensions to form thick retentive prefilter layer on top of normal filter paper.
Enhance filtration speed by coagulating precipitates or suspensions to form thick retentive 'prefilter' layer on top of normal filter paper.
  • Enhance filtration speed by coagulating precipitates or suspensions to form thick retentive “prefilter” layer on top of normal filter paper
TRUSTED_SUSTAINABILITY
Specifica
Descrizione Ash content: 0.015%
Quantità 500 g
Tipo Ashless Clippings