Learn More
Cytiva Whatman™ Polycap Disposable Capsules: 36TC
Click to view available options
Dimensione del poro:
0.2/0.1 μm
0.2/0.2 μm
0.6/0.45 μm
Stile ingresso:
1-1/2 in. Sanitary Inlet
1/4 MNPT
1/4-3/8 in. SB Barb
1/4-3/8 in. SB Tubing
Stile uscita:
1-1/2 in. Sanitary Outlet
1/4-3/8 in. SB Barb
1/4-3/8 in. SB Tubing
Include:
Filling Bell
Filter Capsule Only
Dimensione della confezione:
Pezzo
pezzi 5
Descrizione
- Two-layer filter for long life, high efficiency
- Sterile, nonpyrogenic
- Sterilizing grade filtration capsules
- 0.2μm capsules meet bacterial retention per ASTM F838-83
- Low-protein-binding membrane
- Hydrophilic; low extractables
- With inlet vent
- Filter Media: Two layers polyethersulfone (PES)
Applications:
Sterile filtration of biological and tissue culture solutions. Principally for aqueous solutions.
Specifica
Specifica
| Descrizione | Polycap TC 36 Capsule Filter, 0.65/0.45 μm, sterile, with SB inlet and outlet (1 pc) |
| Materiale (membrana) | PES |
| Dimensione del poro | 0.6/0.45 μm |
| Sterilità | Sterile |
| Area di filtraggio | 440 cm2 |
| Da utilizzare con (applicazione) | Sterile filtration of biological and tissue culture solutions. Principally for aqueous solutions. |
| Materiale alloggiamento | Polypropylene |
| Include | Filter Capsule Only |
| Stile ingresso | 1/4-3/8 in. SB Barb |
| Pressione max | 4.1 bar (60 psi) |
| Vedi altri risultati |