Learn More
Whatman™ Protran™ Nitrocellulose Blotting Membranes: 0.45μm BA85 Protran Sheets and Rolls
Click to view available options
Quantità:
1 Each
Lunghezza (sistema metrico):
15 cm
20 cm
30 cm
Larghezza (sistema metrico):
3 m
Formato:
Roll
Dimensione della confezione:
330g
Pezzo
Descrizione
- Universal membrane suitable for Western, Southern, and Northern blotting
- Hydrophilic, so no prewetting with methanol is required
- High binding capacity and very low background for superior signal-to-noise ratios without stringent washing conditions
- Compatible with all common detection methods, including radioactive, colorimetric, fluorescent, and chemiluminescent
- Provide excellent shelf-life for proteins—proteins maintain molecular recognition activity for up to five years on Protran membranes
- Three pore sizes available—0.45μm is ideal for larger molecular weight samples, 0.2μm ensures binding of samples below 20kDa, 0.1μm is used for molecules below 7kDa
- Also available in circular formats
Specifica
Specifica
| Lunghezza (sistema metrico) | 20 cm |
| Larghezza (sistema metrico) | 3 m |
| Da utilizzare con (applicazione) | Suitable for radioactive, colorimetric, fluorescent, and chemiluminescent detection |
| Capacità di legame | High |
| Quantità | 1 Each |
| Formato | Roll |
| Materiale | Nitrocellulose |
| Dimensione del poro | 0,45 μm |