Learn More
DWK Life Sciences Guarnizioni in alluminio: A strappo
Sfoglia Tutto Wheaton Science Products Prodotti
Click to view available options
Colore:
Blue
Green
Incolore
Red
Dimensione chiusura:
13 mm
20 mm
Dimensione della confezione:
pezzi 1000
Descrizione
Tear-off seals remove completely allowing content to pour from bottle. Wheaton™ Tear-Off Aluminum Seals, Unlined are available in a wide range of sizes and colors.
- Good barrier properties
- Color selection includes natural, red, blue and green
- Chemically resistant to alcohol and acetone, among many others
Specifica
Specifica
| Colore | Blue |
| Materiale | Aluminum |
| Quantità | 1000/Cs. |
| Tipo | Complete Tear-off Cap |
| Dimensione chiusura | 20 mm |