Learn More
DWK Life Sciences Wheaton™ Diazomethane Generators for Gas Chromatography
Sfoglia Tutto Wheaton Science Products Prodotti
Click to view available options
Altezza (sistema metrico):
19 cm
20 cm
Dimensione della confezione:
Pezzo
Descrizione
- Utilizes clear-seal grease-free joints
- «O« ring not required
- Manufactured from low extractable borosilicate glass that conforms to USP Type I and ASMT™ E 438 Type I, Class A requirements
Specifica
Specifica
| Certificazione/Conformità | USP Type I, ASTM E 438 Type I, Class A |
| Altezza (sistema imperiale) | 7.4 in. |
| Altezza (sistema metrico) | 19 cm |
| Include | Plastic standard taper joint clamp, a 13-425 screw cap with cut-out top and a ptfe faced rubber liner |
| Tipo | Diazomethane Generator |