Learn More
DWK Life Sciences Wheaton™ Extended Controlled 20mm LDPE Dropper Tips
Product Code. p-3306208 Shop All Wheaton Science Products Products
missing translation for 'orderingAttributeHoverText'
Colore:
Blue
Green
Natural
Orange
Pink
Purple
Red
White
Yellow
Materiale:
LDPE
Polyethylene
N. per custodia:
100 Cs.
1000 Cs.
missing translation for 'unitSize'
pezzi 100
pezzi 1000
This item is not returnable. View return policy
Product Code. 12436081
missing translation for 'mfr': DWK Life Sciences W242434A
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy
20mm extended controlled dropper tip made of LDPE (Low Density Polyethylene)
  • Extended controlled dropper tip dispenses one drop at a time
  • Average drop size is 44μL ±6μL
  • Case of 100
TRUSTED_SUSTAINABILITY
Specifications
Colore Blue
Materiale Polyethylene
N. per custodia 1000 Cs.
Certificazione/Conformità ISO 9001
Drop Size 41 μL
Product Type Dropper Tip
Da utilizzare con (apparecchiatura) Plastic dropping bottle
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.