Learn More
DWK Life Sciences Wheaton™ Magna-Flex™ Spinner Flasks
Sfoglia Tutto Wheaton Science Products Prodotti
Click to view available options
Capacità (sistema metrico):
125 mL
250 mL
500 mL
1000 mL
3000 mL
6 L
Include:
Flask only
Flask with impeller
Dimensione della confezione:
Pezzo
Descrizione
- Borosilicate glass flasks
- Two large side arms with screw-cap closures
- Flex-type, bulb-shaped glass impeller rotates from a fixed position above the liquid level around an indentation in the base of the flask
- Graduated
- Removable stainless-steel pin immobilizes the impeller during handling or decanting to prevent damage to cells or microcarriers
- Autoclavable flasks and impellers for selected sizes can also be ordered separately
- U.S. Patent No. 3,854,704
Specifica
Specifica
| Capacità (sistema metrico) | 125 mL |
| Materiale | Borosilicate Glass |
| Include | Flask with impeller |
| Giunto del braccio laterale | 33-430 |
| Autoclavabile | Autoclavable |
| Graduated | Yes |
| Dimensione chiusura | 51-440 |