Learn More
3M™ White/Blue Combined SMS Laminate Protective Coveralls
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Dimensione della confezione:
pezzi 20
Descrizione
- Breathable back panel to help improve air ventilation and reduce heat build-up.
- Knit cuffs provide a soft wrist closure for increased comfort compared to elastic closures.
- Elastic waist and ankles for convenience and freedom of movement.
- Two-way zipper with sealable storm flap for convenience and to help provide additional protection.
Specifica
Specifica
| Product Type | General Purpose Coveralls |
| Size | 2X-Large |
| Sesso | Unisex |
| Monouso | Single-use |
| Stile vita | Elastic |
| Stile polso | Knit |
| Stile collo | Con cappuccio |
| Sterilità | Non-sterile |
| Classe di protezione | Category III |
| Altezza (sistema metrico) | 186 to 194 cm |