Learn More
SHOWA™ White General Purpose Gloves, Model B0610
Click to view available options
Size:
6
7
8
9
Dimensione della confezione:
10 paia
Descrizione
- Designed for optimum freedom of movement, the B0610 has a short wrist cuff.
- The uncoated, seamless nylon liner reduces irritation and allergy risks while maintaining dexterity and tactility.
Specifica
Specifica
| Coating Material | Uncoated |
| Colore | White |
| Monouso | Reusable |
| Hand | Ambidestri |
| Laboratory/Cleanroom Use | Sì |
| Latex Free | Sì |
| Materiale | Nylon |
| Product Type | Glove |
| Size | 8 |
| Sterilità | Non-sterile |
| Vedi altri risultati |