Learn More
Heathrow Scientific Bench-Top Keeper™ Biohazard Container
GREENER_CHOICE
Codice prodotto. p-36362 Sfoglia Tutto Heathrow Scientific Prodotti
Click to view available options
Altezza (sistema metrico):
18 cm
25 cm
Lunghezza (sistema metrico):
10 cm
15 cm
Larghezza (sistema metrico):
10 cm
15 cm
Size:
Small
Large
Codice prodotto. 30804786
Marca: Heathrow Scientific BH2002
Economical point-of-use alternative to hard-sided sharps containers for disposal of contaminated Petri dishes, plastic assay vials, analyzer cups and other non-sharp waste
  • Made of sturdy paperboard with an internal plastic lining and webbed corners to contain moderate amounts of fluids
  • Self-locking tops keep contents safely enclosed
  • Small Keepers fold out to 4L x 4W x 7H in. (10 x 10 x 18 cm) and accommodate up to 1600 cc of material
  • Large Keepers fold out to 6L x 6W x 10H in. (15 x 15 x 25 cm) and accommodate up to 5000 cc of material
  • Autoclavable
  • Made in USA
Specifica
Capacità (sistema metrico) 1.4 L
Capacità (sistema imperiale) 1400 cc
Dimensioni (L x P x A) 10 x 10 x 18 cm (4 x 4 x 7 in.)
Monouso Single-use
Altezza (sistema metrico) 18 cm
Lunghezza (sistema metrico) 10 cm
Size Small
Larghezza (sistema metrico) 10 cm
Autoclavabile Autoclavable
Include Self-locking tops
Materiale rivestimento interno Plastic
Da utilizzare con (applicazione) Storing contaminated lab waste, disposal of contaminated petri dishes, plastic assay vials, analyzer cups and other non-sharp waste
Larghezza (sistema imperiale) 4 in.
Lunghezza (sistema imperiale) 4 in.
Altezza (sistema imperiale) 7 in.
Materiale Paperboard
Product Type Benchtop Keeper