Learn More
Agilent BioTek™ Widefield Imaging Accessory Kit
Sfoglia Tutto Agilent BioTek Prodotti
Click to view available options
Da utilizzare con (apparecchiatura):
Agilent BioTek Cytation and Lionheart
Dimensione della confezione:
1 unità
Descrizione
- For use with Cytation Cell Imaging Multimode Readers and Lionheart Automated Microscopes.
- Designed to provide the essential objectives, filters cubes, LEDs and adapters required for use across a broad range of widefield imaging workflows.
- Includes protective case for convenience and ease of use.
Specifica
Specifica
| Product Type | Accessory Kit |
| Da utilizzare con (apparecchiatura) | Agilent BioTek Cytation and Lionheart |
| Sterilità | Non-sterile |