Learn More
Woodsafe™ Woodsafe Sharpsbin Holders
Click to view available options
Da utilizzare con (apparecchiatura):
0.5 L Woodsafe Containers
2 and 3 L Woodsafe Containers
25 L and 50 L Woodsafe Containers
6 L and 12 L Woodsafe Containers
Woodsafe Holders
Product Type:
Holder
String Hook
Table Mount
Trolley
Dimensione della confezione:
Pezzo
pezzi 10
pezzi 100
Descrizione
- Woodsafe™ offer a sustainable clinical waste management solution produced with residue from the forest industry
- Lowers the carbon footprint by more than 60%, proved in full LCA
- Steady holders and trolleys
Specifica
Specifica
| Descrizione | Woodsafe Sharpsbin Holders |
| Da utilizzare con (apparecchiatura) | 2 and 3 L Woodsafe Containers |
| Product Type | Holder |
Lower climate impact and ACT label