Learn More
Cryo-Industrial™ Wrist Fabric Gloves
Click to view available options
Size:
Large
X-Large
Dimensione della confezione:
1 paio
Descrizione
- Protection for hands and arms when working in hazardous, ultra-cold environments down to -200°C
- Gloves are designed to provide a high level of thermal protection and abrasion resistance for heavy-duty applications
- Wrist length
- Made from state-of-the-art materials
- Designed to keep the user dry and to wick moisture away from your hands
- Can be worn in comfort for extended periods of time.
- Multi-layered construction allows for a maximum level of thermal protection, flexibility, and dexterity
Specifica
Specifica
| Colore | Blue |
| Monouso | Reusable |
| Materiale | Fabric |
| Powdered | Powder Free |
| Product Type | Cryogenic Gloves |
| Size | Large |
| Sterilità | Non-sterile |
| Stile | Wrist-length |