Learn More
Abeba™ X-Light Safety Loafer
Click to view available options
Size:
35
36
37
38
39
40
41
42
43
44
45
46
47
48
Dimensione della confezione:
1 paio
Description
- CE, EN ISO 20345:2011, S2, SRC
- Smooth leather
- Black
- Steel toe cap
- Slip resistance PU outsole sole
- Replaceable Active Comfort Insole (art. 351620)
- Antistatic
- Breathable lining with Silver ions
- Instep area with elastic band
Specifications
Specifications
| Tipo di chiusura | Elastic Band |
| Colore | White |
| Materiale | Leather |
| Size | 37 |
| Materiale dello zoccolo | Steel |
| Certifications/Compliance (Europe) | CE, EN ISO 20345:2011, S2, SRC |