Learn More
XBP1S-Specific Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 24868-1-AP-150UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
The XBP1S-specific antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human XBP1S-specific. This antibody recognizes human, mouse antigen. The XBP1S-specific antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifica
| XBP1S-Specific | |
| Polyclonal | |
| Unconjugated | |
| 150 μL | |
| 7494 | |
| IgG |
| Western Blot, Immunohistochemistry | |
| 293 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Mouse |