Description
XK X-linked Kx blood group Polyclonal antibody specifically detects XK X-linked Kx blood group in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
Specifications
| Classification | Polyclonal |
| Dilution | Western Blot, Immunocytochemistry/ Immunofluorescence |
| Gene Alias | Kell blood group precursor (McLeod phenotype), Kell complex 37 kDa component, KX, Kx antigen, membrane transport protein XK, X1k, XK, Kell blood group complex subunit (McLeod syndrome), XKR1XK-related protein 1, X-linked Kx blood group (McLeod syndrome), XRG1 |
| Content And Storage | Store at 4C in the dark. |
| Product Type | Antibody |
| Form | Purified |
| Product Line | Novus |