Learn More
Chemsplash Xtreme 50 SMS T5/6 Coverall with BlueTaped Seams
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Dimensione della confezione:
pezzi 25
Descrizione
Chemsplash Xtreme SMS 50 Coverall
- 50GSM Breathable SMS Fabric
- Fully Taped Seams
- Elasticated 3 Pce Hood
- Elasticated Cuffs & Ankles
- Two Way Zip With Adhesive Zip Flap
- Silicone and Latex Free
- PFAS Free
- Comes without the chest label
Description: The Chemsplash Xtreme SMS 50 Coverall with Taped Seams is made from breathable 50gsm SMS fabric, compliant to Cat III Type 5 and 6. It offers a robust and strong barrier to dry hazards and is especially suitable for prolonged periods of use due to its breathable properties. All seams are bound.
Specifica
Specifica
| Product Type | Hooded Coveralls |
| Size | 3X-Large |
| Sesso | Unisex |
| Materiale | SMS Fabric |
| Colore | Blue |
| Monouso | Single-use |
| Stile vita | Elastic |
| Ankle Style | Elastic |
| Stile polso | Elastic |
| Stile collo | Hooded |
| Vedi altri risultati |