Learn More
Thermo Scientific™ Yersinia Agar/Yersinia Agar Biplate
Descrizione
Isolate and enumerate Yersinia enterocolitica from clinical specimens and food samples after 18 to 24 hours using this bi-plate.
Isolate and enumerate Yersinia enterocolitica from clinical specimens and food samples after 18 to 24 hours using Thermo Scientific™ Oxoid™ Yersinia Agar /Yersinia Agar Biplate. Specifically developed for the optimum growth and recovery of Yersinia enterocolitica. The typical colonies of Yersinia enterocolitica will develop as a red bullseye surrounded by a transparent border and will vary considerably among serotypes in colony size, smoothness and the ratio of the border to center diameter. Save space in automated specimen processing to achieve greater throughput.
Yersinia enterocolitica is a bacterial species in the family Enterobacteriaceae that most often causes enterocolitis, acute diarrhea, terminal ileitis, mesenteric lymphadenitis, and pseudoappendicitis but, if it spreads systemically, can also result in fatal sepsis.
Isolate and enumerate Yersinia enterocolitica in 18-24 hours with Yersinia Agar /Yersinia Agar Biplate.
- Selective: selective for Yersinia enterocolitica
- Easy-to-read: colonies of Yersinia enterocolitica develop as a red bullseye surrounded by a transparent border
- Ready-to-use: convenience of prepared media
- Space saving: bi-plate format
- Increase throughput: fewer plates to process per sample allows greater throughput in automated specimen processors.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
Specifica
Specifica
| Colore | Antique pink, transparent |
| Content And Storage | 2°C to 12°C |
| Descrizione | Yersina Agar / Yersina Agar |
| Formato | Biplate |
| Manufacturing Origin | United Kingdom |
| Product Type | Agar Plate |
| Peso riempimento | 17g + 5% |
| Tipo di imballaggio | Flow Wrapped - Single Layer |
| Quantità | 10 Pack |
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.