Learn More
YSI™ 6052030 Pro Series DO/Conductivity Cables
Codice prodotto. 11927127
Click to view available options
Lunghezza (sistema imperiale):
1181.1 in.
Dimensione della confezione:
Pezzo
Codice prodotto. 11927127
Marca: YSI™ 605203030
Cables available in 4m, 10, 20, 30m lengths
The 6052030 cables ONLY work with the YSI Pro Plus handheld
  • Measure dissolved oxygen, conductivity, salinity, specific conductance, total dissolved solids and temperature
  • Conductivity sensor is built into the cable
  • Automatic salinity compensation for DO readings
  • Cables do NOT come with the DO sensor
TRUSTED_SUSTAINABILITY
Specifica
Da utilizzare con (applicazione) Conductivity, DO, Temp
Include Built-in conductivity sensor and temperature/DO sensor port
Lunghezza (sistema imperiale) 1181.1 in.
Lunghezza (sistema metrico) 30 m