Learn More
YSI™ ProODO Handheld Optical Dissolved Oxygen Meter
Click to view available options
Protezione IP:
IP-67
Dimensione della confezione:
Pezzo
Descrizione
The ProODO instrument meets the demands of true field work.
- No electrodes to clean or solutions to change
- Easy to read graphic, backlit display and keypad for use in any lighting condition
- Expanded DO range of 0-500% with response time of T90 in approximately 25 seconds
- Stores 5000 data sets (sensor data, date, time, site and user defined information)
- USB connectivity for use with Data Manager software
- Non-consumptive luminescent method eliminates stirring
- Impact resistant, IP-67 waterproof case and connectors with rubber non-slip case
- User replaceable sensing element good for approximately one year
NOTE: Probes and cables are not included and must be purchased separately.
Order Info
Meter Kit is also available (09324507).
Warranty
Three-year instrument warranty
Specifica
Specifica
| Include | Data Manager software, the ProComm II communications saddle, and USB cable |
| Tipi di sensore | Optical |
| Risoluzione (temperatura) | 0.1°C |
| Precisione (Temperatura) | ±0.2°C |
| Certificazione/Conformità | RoHS, CE, WEEE, C-Tick, VCCI, FCC, IP67, GLP |
| Protezione IP | IP-67 |
| Peso (sistema metrico) | 0.47 kg |
| Peso (sistema imperiale) | 1.05 lb. |
| Warranty | 3 Years |
| Intervallo (DO) | 0 to 50mg/L |
| Vedi altri risultati |