Learn More
Thermo Scientific Chemicals Yttria Stabilized Zirconium Oxide Cylindrical Crucibles
Click to view available options
Capacità (sistema metrico):
5 mL
10 mL
50 mL
100 mL
200 mL
Diametro (sistema metrico) esterno:
20mm
24mm
37mm
45mm
55mm
Altezza (sistema metrico):
37mm
43mm
72mm
90mm
112mm
Dimensione della confezione:
Pezzo
Descrizione
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifica
Specifica
| Materiale | Zirconium Oxide |
| Quantità | 1 Piece |
| Capacità (sistema metrico) | 200 mL |
| Diametro (sistema metrico) esterno | 55mm |
| Altezza (sistema metrico) | 112mm |
| Product Type | Yttria Stabilized Zirconium Oxide Cylindrical Crucibles |