Learn More
Tocris Bioscience™ Z-DEVD-FMK Caspase-3 Inhibitor
Click to view available options
Quantità:
1 mg
Dimensione della confezione:
1mg
Description
Z-DEVD-FMK Caspase-3 Inhibitor is a cell-permeable, irreversible inhibitor of caspase-3/CPP32. It inhibits tumor cell apoptosis. In vivo, it has a neuroprotective effect in rat hippocampus following seizures. It also significantly reduces post-traumatic apoptosis, and improved neurological recovery, before and after induced traumatic brain injury in rat.
Specifications
Specifications
| Purezza | 95% |
| CAS | 634911-78-7 |
| Content And Storage | Store at -20°C |
| Quantità | 1 mg |
| Target Entry | Caspase-3 |