Learn More
ZBTB2 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 26436-1-AP-150UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
The ZBTB2 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human ZBTB2. This antibody recognizes human antigen. The ZBTB2 antibody has been validated for the following applications: IP, ELISA analysis.
Specifica
| ZBTB2 | |
| 350 μg/mL | |
| Rabbit | |
| Primary | |
| Human | |
| 150 kDa | |
| IgG |
| Immunoprecipitation, ELISA | |
| KIAA1483, ZBTB2, ZNF437 | |
| 150 μL | |
| 57621 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |