missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZIC4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
431.00€ - 618.00€
Specifica
| Antigene | ZIC4 |
|---|---|
| Applicazioni | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classificazione | Polyclonal |
| Coniugato | Unconjugated |
| Specie ospite | Rabbit |
Descrizione
ZIC4 Polyclonal specifically detects ZIC4 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifica
| ZIC4 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| Q8N9L1 | |
| 84107 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: LVMRKRLRLYRNTLKESSSSSGHHGPQLTAASSPSVFPGLHEEPPQASPSRPLNGLLRLGLPGDMYARPEPFPPGPAARSDALAAAAALHGYGGMNLTVNL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| FLJ42609, FLJ45833, Zic family member 4, Zinc finger protein of the cerebellum 4zinc family member 4 protein HZIC4, zinc finger protein ZIC 4 | |
| ZIC4 | |
| IgG | |
| Affinity Purified |
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto