Learn More
Thermo Scientific Chemicals Zirconium Oxide Round Bottom Crucibles, High Form
Click to view available options
Capacità (sistema metrico):
5 mL
10 mL
100 mL
200 mL
Diametro (sistema metrico) esterno:
26mm
30mm
56mm
69mm
Altezza (sistema metrico):
29mm
34mm
68mm
85mm
Dimensione della confezione:
Pezzo
Descrizione
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifica
Specifica
| Materiale | Zirconium Oxide |
| Quantità | 1 Piece |
| Capacità (sistema metrico) | 10 mL |
| Diametro (sistema metrico) esterno | 30mm |
| Altezza (sistema metrico) | 34mm |
| Product Type | Zirconium Oxide Round Bottom Crucibles, High Form |
| Diametro (sistema metrico) base | 20mm |