missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZnT6 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 13526-1-AP-20UL
Questo articolo non è restituibile.
Consulta la politica di reso
Beskrivelse
Zinc functions as a cofactor for numerous enzymes, nuclear factors, and hormones and as an intra- and intercellular signal ion. Members of the zinc transporter (ZNT)/SLC30 subfamily of the cation diffusion facilitator family, such as SLC30A6, permit cellular efflux of zinc.Tekniske data
| ZnT6 | |
| Polyclonal | |
| Unconjugated | |
| SLC30A6 | |
| MST103, MSTP103, SLC30A6, Zinc transporter 6, ZnT 6, ZNT6 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 55676 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q6NXT4 | |
| SLC30A6 | |
| ZnT6 Fusion Protein Ag4403 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion