missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZRANB2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA584178
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Immunogen sequence: TKNFRVSDGD WICPDKKCGN VNFARRTSCN RCGREKTTEA KMMKAGGTEI GKTLAEKSRG LFSANDWQCK TCSNVNWARR S Highest antigen sequence indentity to the following orthologs: Mouse - 100%, Rat - 100%.
Zinc-finger proteins contain DNA-binding domains and have a wide variety of functions, most of which encompass some form of transcriptional activation or repression. ZNF265 (Zinc finger protein 265), also known as ZRANB2 (Zinc finger Ran-binding domain-containing protein 2), ZIS, ZIS1 or ZIS2, is a 330 amino acid protein that belongs to the ZRANB2 family. Localized to the nucleus, ZNF265 functions as a splicing factor that is responsible for alternatively splicing Tra-2 beta (transformer-2 beta) transcripts and is thought to interfere with constitutive 5'-splice selection. ZNF265 contains two RanBP2-type zinc fingers through which it conveys its RNA-binding activity. Two isoforms, designated ZIS-1 and ZIS-2, are expressed due to alternative splicing events. Upon DNA damage, ZIS-2 may be phosphorylated by ATM or ATR.
Specifica
| ZRANB2 | |
| Polyclonal | |
| Unconjugated | |
| Zranb2 | |
| AI227013; DKFZp686J1831; DKFZp686N09117; wu:fa96h03; Zfp265; zgc:56597; zinc finger protein 265; Zinc finger Ran-binding domain-containing protein 2; Zinc finger Ran-binding domain-containing protein 2-like protein; zinc finger RANBP2-type containing 2; zinc finger, RAN-binding domain containing 2; zinc finger, splicing; zinc-finger protein 265; zinc-finger, splicing; ZIS; ZIS1; ZIS2; Znf265; ZRANB2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9406 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry, Immunofluorescence | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O95218 | |
| Zranb2 | |
| Recombinant protein corresponding to Human ZRANB2. Recombinant protein control fragment (Product #RP-101663). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto