CAS RN 10097-07-1
CAS RN 10097-07-1
IUPAC Nome:
—
Sinonimi:
—
Peso molecolare (g/mol):
—
Formula molecolare:
—
InChi Key:
—
SMILES:
—
1
–
3
di
3
risultati
1
N,N’-1,4-Phenylenedi-glycine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
N,N’-1,4-Phenylenedi-glycine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
N,N’-1,4-Phenylenedi-glycine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.