CAS RN 1010733-93-3
CAS RN 1010733-93-3
IUPAC Nome:
—
Sinonimi:
—
Peso molecolare (g/mol):
—
Formula molecolare:
—
InChi Key:
—
SMILES:
—
1
–
3
di
3
risultati
1
[[3,3',3' '-(Phosphinidyne-KappaP)tris[benzenesulfonato]](3-)]-Palladate(3-) Sodium, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
[[3,3',3' '-(Phosphinidyne-KappaP)tris[benzenesulfonato]](3-)]-Palladate(3-) Sodium, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
[[3,3',3' '-(Phosphinidyne-κP)tris[benzenesulfonato]](3-)]-Palladate(3-) Sodium, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.