CAS RN 1014-70-6
CAS RN 1014-70-6
IUPAC Nome:
N2,N4-diethyl-6-(methylsulfanyl)-1,3,5-triazine-2,4-diamine
Sinonimi:
simetryn gy-bon
Peso molecolare (g/mol):
213.3
Formula molecolare:
C8H15N5S
InChi Key:
MGLWZSOBALDPEK-UHFFFAOYSA-N
SMILES:
CCNC1=NC(SC)=NC(NCC)=N1
1
–
2
di
2
risultati
1
Simetryn, MedChemExpress
MedChemExpress Simetryn is a herbicide with the LC50 of 16.9-3.70 mg/L (79.3-17.4 microM.