CAS RN 10500-57-9
CAS RN 10500-57-9
IUPAC Name:
5,6,7,8-tetrahydroquinoline
Sinonimi:
quinoline, 5,6,7,8-tetrahydro-
Peso molecolare (g/mol):
133.19
Formula molecolare:
C9H11N
InChi Key:
YQDGQEKUTLYWJU-UHFFFAOYSA-N
SMILES:
C1CCC2=NC=CC=C2C1
1
–
1
di
1
risultati
1
5,6,7,8-Tetrahydroquinoline, 98%, Thermo Scientific Chemicals
CAS: 10500-57-9 Formula molecolare: C9H11N Molecular Weight (g/mol): 133.19 InChI Key: YQDGQEKUTLYWJU-UHFFFAOYSA-N Sinonimo: 2,3-cyclohexeno pyridine,quinoline, 5,6,7,8-tetrahydro,unii-l786aag56h,5,6,7,8-tetra-hydroquinoline,5,6,7,8-tetrahydro-quinoline,pubchem5891,2,3-cyclohexenopyridine,g00034-watson-int,2,3-cyclohexano pyridine,acmc-2098eh PubChem CID: 66335 IUPAC Name: 5,6,7,8-tetrahydroquinoline SMILES: C1CCC2=C(C1)C=CC=N2