CAS RN 1096708-71-2
CAS RN 1096708-71-2
IUPAC Nome:
—
Sinonimi:
—
Peso molecolare (g/mol):
—
Formula molecolare:
—
InChi Key:
—
SMILES:
—
1
–
1
di
1
risultati
1
TAK-580, MedChemExpress
MedChemExpress TAK-580 (MLN 2480) is an orally active and selective inhibitor of pan-Raf kinase.