CAS RN 1128-76-3
CAS RN 1128-76-3
IUPAC Nome:
ethyl 3-chlorobenzoate
Sinonimi:
ethyl 3-chlorobenzoate
Peso molecolare (g/mol):
184.62
Formula molecolare:
C9H9ClO2
InChi Key:
LVFRSNCBCHABAM-UHFFFAOYSA-N
SMILES:
CCOC(=O)C1=CC=CC(Cl)=C1
1
–
1
di
1
risultati
1
Ethyl 3-chlorobenzoate, 98+%
CAS: 1128-76-3 Formula molecolare: C9H9ClO2 Molecular Weight (g/mol): 184.619 Numero MDL: MFCD00013634 InChI Key: LVFRSNCBCHABAM-UHFFFAOYSA-N Sinonimo: benzoic acid, 3-chloro-, ethyl ester,3-chlorobenzoic acid ethyl ester,ethyl m-chlorobenzoate,benzoic acid, m-chloro-, ethyl ester,3-chloro-benzoic acid ethyl ester,pubchem3704,acmc-1bt4u,ethyl3-chlorobenzoate,ksc495m6f,rarechem al bi 0056 PubChem CID: 70785 IUPAC Name: ethyl 3-chlorobenzoate SMILES: CCOC(=O)C1=CC(=CC=C1)Cl