CAS RN 1197-22-4
IUPAC Nome:
N-phenylmethanesulfonamide
Sinonimi:
N-phenyl methanesulfonamide
Peso molecolare (g/mol):
171.21
Formula molecolare:
C7H9NO2S
InChi Key:
LBTPIFQNEKOAIM-UHFFFAOYSA-N
SMILES:
CS(=O)(=O)NC1=CC=CC=C1
1
–
5
di
5
risultati
2
N-Phenylmethanesulfonamide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
N-Phenylmethanesulfonamide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
N-Phenylmethanesulfonamide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
5
N-Phenylmethanesulfonamide, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.